4-Chinolincarbonsäure
Lieferant: Molekula
|
Achtung
|
Synonyme:
Chinolin-4-carbonsäure
M69509659
MOLEM69509659EA
0
EUR
MOLEM69509659
4-Chinolincarbonsäure
4-Chinolincarbonsäure
Formel:
C₁₀H₇NO₂ Molecular Weight: 173,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006782 CAS-Nummer: 486-74-8 EINECS: 207-640-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...