N-Octyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-8, SB-8)

Lieferant: Molekula
Achtung

Synonyme: 3-(N,N-Dimethyloctylammonio)propanesulphonate inner salt

M69597338
MOLEM69597338EA 0 EUR
MOLEM69597338
N-Octyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-8, SB-8)
N-Octyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-8, SB-8)
Formel: C₁₃H₂₉NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00036912
CAS-Nummer: 15178-76-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR