N-(1-Naphthyl)ethylenediamine Dihydrochlorid

Lieferant: Molekula
Achtung

Synonyme: N-(1-Naphthyl)ethylendiamin Dihydrochlorid, N-(1-Naphthyl)ethylendiammoniumdichlorid, Marshall Reagenz, N-2-Aminoethyl-1-naphthylamin Dihydrochlorid, 2-((Aminoethyl)amino)naphthalin Dihydrochlorid

M69625605
MOLEM69625605EA 0 EUR
MOLEM69625605
N-(1-Naphthyl)ethylenediamine Dihydrochlorid
N-2-Aminoethyl-1-naphthylamin Dihydrochlorid
Formel: C₁₂H₁₆Cl₂N₂
Molecular Weight: 259,18 g/mol
Schmelzpunkt: 188…190 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012556
CAS-Nummer: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR