2-Cyanphenylboronsäure

Lieferant: Molekula
Achtung

Synonyme: Dihydroxy(2-cyanphenyl)boran, 2-Cyanbenzolboronsäure, (2-Cyanphenyl)metaborsäure

M69832405
MOLEM69832405EA 0 EUR
MOLEM69832405
2-Cyanphenylboronsäure
2-Cyanphenylboronsäure
Formel: C₇H₆BNO₂
Molecular Weight: 146,94 g/mol
MDL: MFCD01632208
CAS-Nummer: 138642-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR