4,6-Dichlor-5-nitropyrimidin

Lieferant: Molekula

M69992794
MOLEM69992794EA 0 EUR
MOLEM69992794
4,6-Dichlor-5-nitropyrimidin
4,6-Dichloro-5-nitropyrimidine
Formel: C₄HCl₂N₃O₂
Molecular Weight: 193,98 g/mol
MDL: MFCD00006107
CAS-Nummer: 4316-93-2
EINECS: 224-340-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR