Digitoxin

Lieferant: Molekula

Synonyme: 3β,14β-Dihydroxy-5β-carden-20(22)-olid-3-tridigitoxid

M70068568
MOLEM70068568EA 181 EUR
MOLEM70068568
Digitoxin
Digitoxin
Formel: C₄₁H₆₄O₁₃
Molecular Weight: 764.95 g/mol
Schmelzpunkt: 276
MDL: MFCD00003686
CAS-Nummer: 71-63-6
EINECS: 200-760-5
UN: 2811
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR