5-Chloroxindol

Lieferant: Molekula

Synonyme: 5-Chloro-2-oxindole

M70090951
MOLEM70090951EA 0 EUR
MOLEM70090951
5-Chloroxindol
5-Chloroxindol
Formel: C₈H₆ClNO
Molecular Weight: 167,59 g/mol
MDL: MFCD00191927
CAS-Nummer: 17630-75-0
EINECS: 412-200-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR