2-Amino-5-brom-3-nitropyridin
Lieferant: Molekula
|
Achtung
|
Synonyme:
5-Brom-3-nitropyridin-2-amin, 5-Brom-3-nitro-2-pyridylamin
M70222392
MOLEM70222392EA
0
EUR
MOLEM70222392
2-Amino-5-brom-3-nitropyridin
2-Amino-5-brom-3-nitropyridin
Formel:
C₅H₄BrN₃O₂ Molecular Weight: 218,01 g/mol Schmelzpunkt: 215…216 °C |
MDL:
MFCD00047441 CAS-Nummer: 6945-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...