3-Bromphenetol

Lieferant: Molekula

Synonyme: 1-Brom-3-ethoxybenzol, (3-Bromphenyl)ethylether

M70318195
MOLEM70318195EA 0 EUR
MOLEM70318195
3-Bromphenetol
3-Bromphenetol
Formel: C₈H₉BrO
Molecular Weight: 201,06 g/mol
MDL: MFCD00274287
CAS-Nummer: 2655-84-7
EINECS: 220-186-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR