3-Chlor-4-fluortoluol

Lieferant: Molekula
Gefahr

Synonyme: 1-Chlor-2-fluor-5-methylbenzol

M70394779
MOLEM70394779EA 0 EUR
MOLEM70394779
3-Chlor-4-fluortoluol
3-Chlor-4-fluortoluol
Formel: C₇H₆ClF
Molecular Weight: 144.58 g/mol
MDL: MFCD00079730
CAS-Nummer: 1513-25-3
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR