1-Methyl-4-piperidon

Lieferant: Molekula

M71038740
MOLEM71038740EA 0 EUR
MOLEM71038740
1-Methyl-4-piperidon
1-Methyl-4-piperidon
Formel: C₆H₁₁NO
MDL: MFCD00006191
CAS-Nummer: 1445-73-4
EINECS: 215-895-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR