DL-Tyrosin
Lieferant: Molekula
Synonyme:
4-Hydroxyphenylalanin, 2-Amino-3-(4-hydroxyphenyl)propionsäure, H-DL-Tyr-OH, 3-(p-Hydroxyphenyl)-DL-alanin
M71128222
MOLEM71128222EA
0
EUR
MOLEM71128222
DL-Tyrosin
DL-Tyrosin
Formel:
C₉H₁₁NO₃ Siedepunkt: 378 °C (1013 hPa) Schmelzpunkt: 285 °C |
MDL:
MFCD00063074 CAS-Nummer: 556-03-6 EINECS: 209-113-1 Merck Index: 12,09970 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...