3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-on
Lieferant: Molekula
Synonyme:
1,4-Cyclohexanedione mono(2,2-dimethyltrimethylene ketal)
M71149744
MOLEM71149744EA
0
EUR
MOLEM71149744
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-on
3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-on
Formel:
C₁₁H₁₈O₃ Molecular Weight: 198,26 g/mol |
MDL:
MFCD00006652 CAS-Nummer: 69225-59-8 EINECS: 273-918-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...