2-(4-Iodphenyl)-3-(4-nitrophenyl)-5-phenyltetrazoliumchlorid

Lieferant: Molekula
Achtung

Synonyme: 3-(4-Iodphenyl)-2-(4-nitrophenyl)-5-phenyltetrazoliumchlorid, 2-(4-Iodphenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazoliumchlorid, INT, Iodnitrotetrazoliumchlorid, 3-(4-Iodphenyl)-2-(4-nitrophenyl)-5-phenyl-2H-tetrazoliumchlorid

M71641895
MOLEM71641895EA 0 EUR
MOLEM71641895
2-(4-Iodphenyl)-3-(4-nitrophenyl)-5-phenyltetrazoliumchlorid
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride
Formel: C₁₉H₁₃ClIN₅O₂
Molecular Weight: 505,7 g/mol
Schmelzpunkt: 227…229 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00011961
CAS-Nummer: 146-68-9
EINECS: 205-676-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR