Acetylcholiniodid
Lieferant: Molekula
|
Achtung
|
Synonyme:
(2-Acetoxyethyl)trimethylammoniumiodid, Acetylcholiniumiodid
M71763567
MOLEM71763567EA
0
EUR
MOLEM71763567
Acetylcholiniodid
Acetylcholiniodid
Formel:
H₃CC(O)O(CH₂)₂N(CH₃)₃I Molecular Weight: 273,11 g/mol Schmelzpunkt: 163…165 °C Dichte: 1,67 g/cm³ (20 °C) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00011815 CAS-Nummer: 2260-50-6 EINECS: 218-862-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...