Decyl-β-D-glucopyranosid

Lieferant: Molekula
Achtung

M72640496
MOLEM72640496EA 0 EUR
MOLEM72640496
Decyl-β-D-glucopyranosid
Decyl-β-D-glucopyranosid
Formel: C₁₆H₃₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00063297
CAS-Nummer: 58846-77-8
EINECS: 261-469-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR