2-Hexinsäuremethylester

Lieferant: Molekula

M72661626
MOLEM72661626EA 0 EUR
MOLEM72661626
2-Hexinsäuremethylester
2-Hexynoic acid methyl ester
Formel: C₇H₁₀O₂
Molecular Weight: 126,16 g/mol
MDL: MFCD00041642
CAS-Nummer: 18937-79-6
EINECS: 242-690-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR