2-Aminophenylboronsäure

Lieferant: Molekula

Synonyme: 2-Aminobenzolboronsäure, Dihydroxy(2-aminophenyl)boran, (2-Aminophenyl)metaborsäure

M72836250
MOLEM72836250EA 0 EUR
MOLEM72836250
2-Aminophenylboronsäure
2-Aminophenylboronsäure
Formel: C₆H₈BNO₂
Molecular Weight: 136,95 g/mol
MDL: MFCD01074645
CAS-Nummer: 5570-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR