2-Bromchinolin

Lieferant: Molekula
Achtung

M73168689
MOLEM73168689EA 0 EUR
MOLEM73168689
2-Bromchinolin
2-Bromoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,06 g/mol
MDL: MFCD00234480
CAS-Nummer: 2005-43-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR