Benzyltriethylammoniumtetrafluoroborat
Lieferant: Molekula
|
Achtung
|
Synonyme:
N-Benzyl-N,N-diethylethanaminium tetrafluoroborate
M73318216
MOLEM73318216EA
0
EUR
MOLEM73318216
Benzyltriethylammoniumtetrafluoroborat
Benzyltriethylammoniumtetrafluoroborat
Formel:
C₁₃H₂₂BF₄N Molecular Weight: 279,13 g/mol Schmelzpunkt: 115…116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011823 CAS-Nummer: 77794-93-5 EINECS: 278-768-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...