2-Thenoesäurehydrazid
Lieferant: Molekula
Synonyme:
2-Thiophencarbonsäurehydrazid, Thiophen-2-carbonsäurehydrazid, Thiophen-2-carbohydrazid
M73362117
MOLEM73362117EA
0
EUR
MOLEM73362117
2-Thenoesäurehydrazid
2-Thenoesäurehydrazid
Formel:
C₅H₆N₂OS Molecular Weight: 142,18 g/mol Schmelzpunkt: 142…143 °C |
MDL:
MFCD00005435 CAS-Nummer: 2361-27-5 EINECS: 219-107-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...