7-Hydroxy-2-tetralon

Lieferant: Molekula

Synonyme: 7-Hydroxy-3,4-dihydro-1H-naphthalen-2-one

M73492513
MOLEM73492513EA 0 EUR
MOLEM73492513
7-Hydroxy-2-tetralon
7-Hydroxy-2-tetralon
Formel: C₁₀H₁₀O₂
Molecular Weight: 162.19 g/mol
MDL: MFCD04038657
CAS-Nummer: 37827-68-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR