N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-14, SB-14)

Lieferant: Molekula

Synonyme: 3-(N,N-Dimethylmyristylammonio)propanesulphonate, Tetradecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner Salt

M73580995
MOLEM73580995EA 0 EUR
MOLEM73580995
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-14, SB-14)
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-14, SB-14)
Formel: C₁₉H₄₁NO₃S
Molecular Weight: 363,5988 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036910
CAS-Nummer: 14933-09-6
EINECS: 239-003-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR