6-Amino-1-hexanol
Lieferant: Molekula
|
Achtung
|
Synonyme:
6-Aminohexan-1-ol
M73615851
MOLEM73615851EA
0
EUR
MOLEM73615851
6-Amino-1-hexanol
6-Amino-1-hexanol
Formel:
C₆H₁₅NO Molecular Weight: 117,19 g/mol Siedepunkt: 227 °C (1013 hPa) Schmelzpunkt: 53…55 °C |
MDL:
MFCD00008241 CAS-Nummer: 4048-33-3 EINECS: 223-748-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...