3-Picolylchlorid Hydrochlorid

Lieferant: Molekula

Synonyme: 3-(Chloromethyl)-pyridinium chloride, 3-(Chloromethyl)pyridine hydrochloride

M73820015
MOLEM73820015EA 0 EUR
MOLEM73820015
3-Picolylchlorid Hydrochlorid
3-Picolylchlorid Hydrochlorid
Formel: C₆H₇Cl₂N
Molecular Weight: 164,03 g/mol
MDL: MFCD00012818
CAS-Nummer: 6959-48-4
EINECS: 230-150-4
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR