(S)-(+)-2-Fluor-α-methyl-4-biphenylessigsäure
Lieferant: Molekula
Synonyme:
(S)-Flurbiprofen, (S)-2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid
M74141640
MOLEM74141640EA
0
EUR
MOLEM74141640
(S)-(+)-2-Fluor-α-methyl-4-biphenylessigsäure
(S)-(+)-2-Fluor-α-methyl-4-biphenylessigsäure
Formel:
C₁₅H₁₃FO₂ Molecular Weight: 244,260 g/mol |
MDL:
MFCD00866152 CAS-Nummer: 51543-39-6 EINECS: 257-263-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...