3-Chlor-4-fluorbenzoesäure

Lieferant: Molekula

M74233060
MOLEM74233060EA 0 EUR
MOLEM74233060
3-Chlor-4-fluorbenzoesäure
3-Chloro-4-fluorobenzoic acid
Formel: C₇H₄ClFO₂
Molecular Weight: 174,56 g/mol
Siedepunkt: 273 °C (1013 hPa)
Schmelzpunkt: 136 °C
MDL: MFCD00042477
CAS-Nummer: 403-16-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR