N-(4-Brombutyl)phthalimid
Lieferant: Molekula
Synonyme:
N-(4-Brombutyl)-1,2-benzoldicarbonsäureimid, N-(4-Brombutyl)phthalsäureimid
M74614667
MOLEM74614667EA
0
EUR
MOLEM74614667
N-(4-Brombutyl)phthalimid
N-(4-Brombutyl)phthalimid
Formel:
C₁₂H₁₂BrNO₂ Schmelzpunkt: 77…80 °C |
MDL:
MFCD00005905 CAS-Nummer: 5394-18-3 EINECS: 226-401-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...