5-Chinolincarbonsäure

Lieferant: Molekula
Achtung

M74870394
MOLEM74870394EA 0 EUR
MOLEM74870394
5-Chinolincarbonsäure
5-Chinolincarbonsäure
Formel: C₁₀H₇NO₂
Molecular Weight: 173,17 g/mol
MDL: MFCD00047608
CAS-Nummer: 7250-53-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR