2,6-Pyridindimethanol

Lieferant: Molekula

Synonyme: 2,6-Bis(hydroxymethyl)pyridine

M74966840
MOLEM74966840EA 0 EUR
MOLEM74966840
2,6-Pyridindimethanol
2,6-Pyridindimethanol
Formel: C₇H₉NO₂
Molecular Weight: 139,15 g/mol
MDL: MFCD00006351
CAS-Nummer: 1195-59-1
EINECS: 214-803-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR