6-Bromchinolin

Lieferant: Molekula

M75339528
MOLEM75339528EA 0 EUR
MOLEM75339528
6-Bromchinolin
6-Bromoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,06 g/mol
MDL: MFCD00024023
CAS-Nummer: 5332-25-2
EINECS: 226-238-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR