2-Aminonicotinsäure

Lieferant: Molekula

Synonyme: 2-Aminopyridin-3-carbonsäure, 2-Amino-3-pyridincarbonsäure

M75348842
MOLEM75348842EA 0 EUR
MOLEM75348842
2-Aminonicotinsäure
2-Aminonicotinsäure
Formel: C₆H₆N₂O₂
Molecular Weight: 138,13 g/mol
Schmelzpunkt: 297 °C
Dichte: 1,545 g/cm³ (20 °C)
MDL: MFCD00006318
CAS-Nummer: 5345-47-1
EINECS: 226-296-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR