4-Tolylboronsäure

Lieferant: Molekula

Synonyme: (4-Methylphenyl)metaborsäure, 4-Methylphenylboronsäure, 4-Methylbenzolboronsäure, Dihydroxy(4-methylphenyl)boran

M75586172
MOLEM75586172EA 0 EUR
MOLEM75586172
4-Tolylboronsäure
4-Tolylboronsäure
Formel: C₇H₉BO₂
Molecular Weight: 135,96 g/mol
Schmelzpunkt: 260 °C
MDL: MFCD00039138
CAS-Nummer: 5720-05-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR