4-Chlor-2-fluorbenzoesäure

Lieferant: Molekula
Achtung

M75667669
MOLEM75667669EA 0 EUR
MOLEM75667669
4-Chlor-2-fluorbenzoesäure
4-Chloro-2-fluorobenzoic acid
Formel: C₇H₄ClFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00042468
CAS-Nummer: 446-30-0
EINECS: 207-162-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR