N-Acetylglycin (Ac-Gly-OH)

Lieferant: Molekula

Synonyme: Ac-Gly-OH, N-Acetylglycine, Acetamidoacetic acid, Aceturic acid

M76007738
MOLEM76007738EA 0 EUR
MOLEM76007738
N-Acetylglycin (Ac-Gly-OH)
N-Acetylglycin (Ac-Gly-OH)
Formel: C₄H₇NO₃
Molecular Weight: 117,1 g/mol
MDL: MFCD00004275
CAS-Nummer: 543-24-8
EINECS: 208-839-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR