α-Brom-m-toluonitril

Lieferant: Molekula

Synonyme: m-(Brommethyl)benzonitril, 3-(Brommethyl)benzoesäurenitril, 3-(Brommethyl)benzonitril

M76614398
MOLEM76614398EA 0 EUR
MOLEM76614398
α-Brom-m-toluonitril
α-Brom-m-toluonitril
Formel: C₈H₆BrN
Molecular Weight: 196,05 g/mol
Siedepunkt: 130 °C (4 mmHg)
Schmelzpunkt: 90…93 °C
MDL: MFCD00001809
CAS-Nummer: 28188-41-2
EINECS: 248-890-1
UN: 2923
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR