2-Mercaptoethansulfonsäure Natriumsalz (Mesna)

Lieferant: Molekula

Synonyme: Sodium 2-mercaptoethanesulphonate

M76880547
MOLEM76880547EA 0 EUR
MOLEM76880547
2-Mercaptoethansulfonsäure Natriumsalz (Mesna)
2-Mercaptoethansulfonsäure Natriumsalz (Mesna)
Formel: C₂H₅NaO₃S₂
Molecular Weight: 164.18 g/mol
MDL: MFCD00007535
CAS-Nummer: 19767-45-4
EINECS: 243-285-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR