4-Fluorphenylboronsäure
Lieferant: Molekula
|
Achtung
|
Synonyme:
Dihydroxy(4-fluorphenyl)boran, 4-Fluorbenzolboronsäure, (4-Fluorphenyl)metaborsäure
M76942805
MOLEM76942805EA
0
EUR
MOLEM76942805
4-Fluorphenylboronsäure
4-Fluorphenylboronsäure
Formel:
C₆H₆BFO₂ Molecular Weight: 139,92 g/mol Schmelzpunkt: 263…265 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039136 CAS-Nummer: 1765-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...