2-(4-Nitrophenyl)ethanol

Lieferant: Molekula

Synonyme: 4-Nitrophenethyl alcohol, p-Nitrophenethyl Alcohol

M77045227
MOLEM77045227EA 0 EUR
MOLEM77045227
2-(4-Nitrophenyl)ethanol
2-(4-Nitrophenyl)ethanol
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
MDL: MFCD00010202
CAS-Nummer: 100-27-6
EINECS: 202-835-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR