2-Ethoxy-1-naphthoesäure

Lieferant: Molekula

Synonyme: 2-Ethoxynaphthoic acid

M77059512
MOLEM77059512EA 0 EUR
MOLEM77059512
2-Ethoxy-1-naphthoesäure
2-Ethoxy-1-naphthoesäure
Formel: C₁₃H₁₂O₃
Molecular Weight: 216,24 g/mol
MDL: MFCD00004008
CAS-Nummer: 2224-00-2
EINECS: 218-745-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR