Methyl-2-oxocyclopentancarboxylat

Lieferant: Molekula

Synonyme: Methyl 2-cyclopentanonecarboxylate

M77223630
MOLEM77223630EA 0 EUR
MOLEM77223630
Methyl-2-oxocyclopentancarboxylat
Methyl-2-oxocyclopentancarboxylat
Formel: C₇H₁₀O₃
Molecular Weight: 142,16 g/mol
MDL: MFCD00001411
CAS-Nummer: 10472-24-9
EINECS: 233-962-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR