1,5-Pentamethylen-1H-tetrazol

Lieferant: Molekula

Synonyme: Pentylenetetrazole

M77238418
MOLEM77238418EA 0 EUR
MOLEM77238418
1,5-Pentamethylen-1H-tetrazol
1,5-Pentamethylen-1H-tetrazol
Formel: C₆H₁₀N₄
Molecular Weight: 138,17 g/mol
MDL: MFCD00005939
CAS-Nummer: 54-95-5
EINECS: 200-219-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR