Glycin-tert-butylester Hydrochlorid (H-Gly-OtBu.HCl)

Lieferant: Molekula

Synonyme: H-Gly-OtBu.HCl, Glycine tert-butyl ester hydrochloride, H-Gly-OtBu·HCl

M77341112
MOLEM77341112EA 0 EUR
MOLEM77341112
Glycin-tert-butylester Hydrochlorid (H-Gly-OtBu.HCl)
Glycin-tert-butylester Hydrochlorid (H-Gly-OtBu.HCl)
Formel: C₆H₁₄ClNO₂
Molecular Weight: 167,64 g/mol
MDL: MFCD00058255
CAS-Nummer: 27532-96-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR