2-Desoxy-L-ribose

Lieferant: Molekula

Synonyme: 2-Desoxy-L(+)-erythropentose, 2-Desoxy-L(+)-arabinose

M77942583
MOLEM77942583EA 0 EUR
MOLEM77942583
2-Desoxy-L-ribose
2-Desoxy-L-ribose
Formel: C₅H₁₀O₄
Molecular Weight: 134,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00132941
CAS-Nummer: 18546-37-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR