Acriflavin
Lieferant: Molekula
|
Gefahr
|
Synonyme:
3,6-Diamino-10-methylacridinium chloride, 3,6-Diamino-10-methylacridinium chloride-3,6-acridinediamine mix, Acriflavinium chloride 3,6-Acridinediamine mix, 3,6-Diamino-10-methylacridinium chloride mixt. with 3,6-diaminoacridine (proflavine), Euflavine, Trypaflavine Neutral, Euflavine, Acriflavine base
M78060746
MOLEM78060746EA
0
EUR
MOLEM78060746
Acriflavin
Acriflavin
Formel:
C₂₇H₂₅ClN₆ Molecular Weight: 468,9806 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064307 CAS-Nummer: 8048-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...