Methyltetrahydropyran-4-carboxylat
Lieferant: Molekula
Synonyme:
Methyl tetrahydro-2H-pyran-4-carboxylate
M78224733
MOLEM78224733EA
0
EUR
MOLEM78224733
Methyltetrahydropyran-4-carboxylat
Methyltetrahydropyran-4-carboxylat
Formel:
C₇H₁₂O₃ Molecular Weight: 144,1684 g/mol |
MDL:
MFCD01075170 CAS-Nummer: 110238-91-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...