3,3'-Diaminodiphenylsulfon

Lieferant: Molekula

Synonyme: 3,3'-Sulfonyldianilin, DADPS, 3-Aminophenylsulfon, Bis(3-aminophenyl)sulfon

M78282256
MOLEM78282256EA 0 EUR
MOLEM78282256
3,3'-Diaminodiphenylsulfon
3,3'-Diaminodiphenylsulfon
Formel: C₁₂H₁₂N₂O₂S
MDL: MFCD00007792
CAS-Nummer: 599-61-1
EINECS: 209-967-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR