Biebricher Scharlachrot

Lieferant: Molekula

Synonyme: CI 26905

M78385699
MOLEM78385699EA 0 EUR
MOLEM78385699
Biebricher Scharlachrot
Biebricher Scharlachrot
Formel: C₂₂H₁₄N₄Na₂O₇S₂
Molecular Weight: 556,49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003891
CAS-Nummer: 4196-99-0
EINECS: 224-084-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR