2-Hydroxy-1-naphthaldehyd

Lieferant: Molekula

Synonyme: 2-Hydroxy-α-naphthaldehyde

M78626682
MOLEM78626682EA 0 EUR
MOLEM78626682
2-Hydroxy-1-naphthaldehyd
2-Hydroxy-1-naphthaldehyd
Formel: C₁₁H₈O₂
Molecular Weight: 172,18 g/mol
MDL: MFCD00004005
CAS-Nummer: 708-06-5
EINECS: 211-902-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR