Hydrocortison
Lieferant: Molekula
Synonyme:
Cortisol, 11β,17α,21-Trihydroxy-4-pregnen-3,20-dion, 11β,17α,21-Trihydroxypregn-4-en-3,20-dion, 11β-Hydroxycortison
M78977397
MOLEM78977397EA
0
EUR
MOLEM78977397
Hydrocortison
Hydrocortison
Formel:
C₂₁H₃₀O₅ Molecular Weight: 362,47 g/mol Siedepunkt: 567 °C (1013 hPa) Schmelzpunkt: 202…204 °C Dichte: 1,4 g/cm³ (20 °C) |
MDL:
MFCD00011654 CAS-Nummer: 50-23-7 EINECS: 200-020-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...